This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PSMD8 SIRNA Assays ›
Gene Symbol:
Psmd8
Gene Name:
proteasome (prosome, macropain) 26S subunit, non-ATPase, 8
Gene Aliases:
6720456J22Rik, AA407360, AL033291, AL033322, AL033323, C76433
Chromosome Location:
Chr.7: 29174187 - 29180673 on Build GRCm38
UniGene ID:
Mm.273152
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 80733

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmd8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026545.3 NP_080821.3 4 699
GenBank mRNA AK002921.1 - 4 538
AK003436.1 - 4 540
AK020132.1 - 4 558
AK027973.1 - 4 503
AK051257.1 - 4 551
AK150493.1 - 4 542
AK150982.1 - 4 506
AK153329.1 - 4 540
AK159216.1 - 4 508
AK159795.1 - 4 524
AK159926.1 - 4 534
AK166718.1 - 4 533
AK167948.1 - 4 508
AK168886.1 - 4 507
BC004075.1 - 4 517 3591796
BC005717.1 - 4 427 2609781

Back To Top

More Information


Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
regulation of protein stability
positive regulation of protein targeting to mitochondrion
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nqfhd:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline