This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NPC1 SIRNA Assays ›
Gene Symbol:
NPC1
Gene Name:
NPC intracellular cholesterol transporter 1
Gene Aliases:
NPC
Chromosome Location:
Chr.18: 23506184 - 23586617 on Build GRCh38
UniGene ID:
Hs.464779
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 8092

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NPC1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000271.4 NP_000262.2 2 441
XM_005258277.1 XP_005258334.1 2 330
XM_005258278.4 XP_005258335.1 2 330
XM_005258279.2 XP_005258336.1 2 330
XM_006722479.3 XP_006722542.1 2 330
XM_011526015.1 XP_011524317.1 2 578
XM_017025784.1 XP_016881273.1 2 330
XM_017025785.1 XP_016881274.1 2 330
XM_017025786.1 XP_016881275.1 2 330
XM_017025787.1 XP_016881276.1 2 330
GenBank mRNA AB209048.1 - 2 202
AB971140.1 - 2 167
AF002020.1 - 2 290
BC063302.1 - 2 264 30340517
BP217319.1 - 2 330

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
endocytosis
autophagy
lysosomal transport
signal transduction
adult walking behavior
cholesterol metabolic process
bile acid metabolic process
negative regulation of macroautophagy
cholesterol transport
membrane raft organization
cholesterol efflux
response to drug
cholesterol homeostasis
response to cadmium ion
viral entry into host cell
negative regulation of cell death
cellular response to steroid hormone stimulus
cellular response to low-density lipoprotein particle stimulus
establishment of protein localization to membrane
positive regulation of cholesterol homeostasis
virus receptor activity
receptor activity
transmembrane signaling receptor activity
protein binding
sterol transporter activity
cholesterol binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0