siRNA Details
| Gene Symbol: Tspan11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026743.3 | NP_081019.1 | 5, 6 | 729 | ||
| GenBank mRNA | AK003691.1 | - | 5, 6 | 581 | ||
| AK028808.1 | - | 5, 6 | 620 | |||
| AK053788.1 | - | 5, 6 | 716 | |||
| AK087294.1 | - | 5, 6 | 744 | |||
| AK090022.1 | - | 5, 6 | 729 | |||
| BC057639.1 | - | 5, 6 | 571 | 4218418 | ||

Tspan11
Genomic Map