This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM114A1 SIRNA Assays ›
Gene Symbol:
Fam114a1
Gene Name:
family with sequence similarity 114, member A1
Gene Aliases:
1190001N04Rik, 9130005N14Rik, Noxp20
Chromosome Location:
Chr.5: 64970075 - 65041901 on Build GRCm38
UniGene ID:
Mm.258545
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 81182

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam114a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026667.3 NP_080943.3 7, 8 1034
GenBank mRNA AK007950.1 - 7, 8 992
AK014725.1 - 7, 8 1004
AK017336.1 - 7, 8 994
AK020262.1 - 7, 8 995
AK030360.1 - 7, 8 1035
AM162548.1 - 7, 8 1038
BC027235.1 - 7, 8 997 3603083
BC110359.1 - 7, 8 1035 5325099

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-j9mnl:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline