This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1810011O10RIK SIRNA Assays ›
Gene Symbol:
1810011O10Rik
Gene Name:
RIKEN cDNA 1810011O10 gene
Gene Aliases:
1110065B09Rik, AW121743, AW321058
Chromosome Location:
Chr.8: 24437616 - 24438946 on Build GRCm38
UniGene ID:
Mm.25775
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 81282

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1810011O10Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026931.2 NP_081207.1 1 216
GenBank mRNA AK007441.1 - 1 217
BC016562.1 - 1 173 3154083

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
regulation of hemopoiesis
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline