siRNA Details
| Gene Symbol: Stoml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026942.3 | NP_081218.3 | 4 | 591 | ||
| GenBank mRNA | AK007508.1 | - | 2 | 260 | ||
| AK045820.1 | - | 4 | 563 | |||
| AK148324.1 | - | 490 | ||||
| BC017536.1 | - | 5 | 843 | |||
| BC037074.1 | - | 4 | 521 | 5037636 | ||

Stoml1
Genomic Map