This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other UFSP1 SIRNA Assays ›
Gene Symbol:
Ufsp1
Gene Name:
UFM1-specific peptidase 1
Gene Aliases:
2700038N03Rik, D5Ertd655e
Chromosome Location:
Chr.5: 137294669 - 137295664 on Build GRCm38
UniGene ID:
Mm.124666
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 81778

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ufsp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027356.2 NP_081632.1 1 229
GenBank mRNA AK012343.1 - 1 230
BC016577.1 - 1 238 3584484
BC087958.1 - 1 250 6751787

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006249198.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
peptidase activity
cysteine-type peptidase activity
hydrolase activity
thiolester hydrolase activity
UFM1 hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-w8hc8:80/100.66.77.245:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0