This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PCBD1 SIRNA Assays ›
Gene Symbol:
PCBD1
Gene Name:
pterin-4 alpha-carbinolamine dehydratase 1
Gene Aliases:
DCOH, PCBD, PCD, PHS
Chromosome Location:
Chr.10: 70882280 - 70888786 on Build GRCh38
UniGene ID:
Hs.3192
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 8191

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PCBD1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000281.3 NP_000272.1 4 748
NM_001289797.1 NP_001276726.1 4 894
GenBank mRNA AF082858.1 - 4 569
BC006324.2 - 4 513 4138350
L41559.1 - 4 516
M83742.1 - 3 497

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
L-phenylalanine catabolic process
tetrahydrobiopterin biosynthetic process
regulation of protein homodimerization activity
positive regulation of transcription, DNA-templated
protein homotetramerization
protein heterooligomerization
oxidation-reduction process
transcription coactivator activity
phenylalanine 4-monooxygenase activity
protein binding
4-alpha-hydroxytetrahydrobiopterin dehydratase activity
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline