This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PSG21 SIRNA Assays ›
Gene Symbol:
Psg21
Gene Name:
pregnancy-specific glycoprotein 21
Gene Aliases:
1600019C01Rik, 1600025N01Rik, 1600026N13Rik, cea8
Chromosome Location:
Chr.7: 18646654 - 18656725 on Build GRCm38
UniGene ID:
Mm.326858
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 81978

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psg21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027403.4 NP_081679.2 5 1642
GenBank mRNA AK005492.1 - 5 1640
AK005545.1 - 5 1285
BC100324.1 - 5 1619 30715195
BC100325.1 - 5 1619 30787148

Back To Top

More Information


Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline