This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ATP6AP2 SIRNA Assays ›
Gene Symbol:
Atp6ap2
Gene Name:
ATPase, H+ transporting, lysosomal accessory protein 2
Gene Aliases:
(P)RR, 5730403E06Rik, APT6M8-9, ATP6M8-9, Atp6ip2, M8-9
Chromosome Location:
Chr.X: 12587759 - 12617051 on Build GRCm38
UniGene ID:
Mm.25148
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 81992

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Atp6ap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027439.4 NP_081715.1 9 1643
GenBank mRNA AK017482.1 - 9 1575
AK029405.1 - 9 1547
AK076495.1 - 9 1588
AK168139.1 - 9 1600
BC014706.1 - 9 1514 4457079

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: BC079339.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

angiotensin maturation
rostrocaudal neural tube patterning
positive regulation of Wnt signaling pathway
positive regulation of transforming growth factor beta1 production
regulation of MAPK cascade
eye pigmentation
head morphogenesis
aspartic-type endopeptidase activity
receptor activity
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline