siRNA Details
| Gene Symbol: Ubiad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027873.2 | NP_082149.1 | 1, 2 | 832 | ||
| GenBank mRNA | AK004550.1 | - | 1, 2 | 832 | ||
| AK078442.2 | - | 1, 2 | 833 | |||
| AK178750.1 | - | 1, 2 | 79 | |||
| AK188039.1 | - | 1, 2 | 79 | |||
| BC015303.1 | - | 1, 2 | 540 | 4241204 | ||
| BC071203.1 | - | 1, 2 | 821 | 3667534 | ||

Ubiad1
Genomic Map