siRNA Details
| Gene Symbol: Nkiras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028024.2 | NP_082300.1 | 3 | 196 | ||
| GenBank mRNA | AK010371.1 | - | 3 | 196 | ||
| AK015918.1 | - | 201 | ||||
| AK052645.1 | - | 3 | 196 | |||
| AK083475.1 | - | 3 | 239 | |||
| AK149696.1 | - | 3 | 340 | |||
| AK169878.1 | - | 3 | 221 | |||
| BC013469.1 | - | 2 | 149 | 4219892 | ||
| BC024398.1 | - | 2 | 139 | 5003001 | ||

Nkiras2
Genomic Map