This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700001F09RIK SIRNA Assays ›
Gene Symbol:
1700001F09Rik BC061237 Gm10375 Gm10377 Gm5800
Gene Name:
RIKEN cDNA 1700001F09 gene
cDNA sequence BC061237
predicted gene 10375
predicted gene 10377
predicted gene 5800
Gene Aliases:
Gm10377
-
ENSMUSG00000072602
ENSMUSG00000072606
EG545047
Chromosome Location:
Chr.14: 43342384 - 43347811 on Build GRCm38
Chromosome Location:
Chr.14: 44500122 - 44506345 on Build GRCm38
Chromosome Location:
Chr.14: 43602627 - 43608013 on Build GRCm38
Chromosome Location:
Chr.14: 42792568 - 42797994 on Build GRCm38
Chromosome Location:
Chr.14: 51711644 - 51717132 on Build GRCm38
UniGene ID:
Mm.335855
Mm.293026
Mm.429750
Mm.429109
Mm.425915
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
-
siRNA ID 82529

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700001F09Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027940.2 NP_082216.1 4, 5 543
GenBank mRNA AK005587.1 - 4, 5 544
BC061060.1 - 4, 5 525 6770724
Gene Symbol: BC061237
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198677.1 NP_941079.1 4, 5 558
GenBank mRNA BC061237.1 - 4, 5 558 6705671
Gene Symbol: Gm10375
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098269.2 NP_001091739.1 4, 5 495
GenBank mRNA BB679674.1 - 1, 2 69
BC089536.1 - 4, 5 495 6742411
Gene Symbol: Gm10377
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244671.1 NP_001231600.1 4, 5 506
GenBank mRNA BC049548.1 - 4, 5 506
Gene Symbol: Gm5800
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034102.2 NP_001029274.1 4, 5 512
GenBank mRNA BC049651.1 - 4, 5 512
BC100479.1 - 4, 5 512 30917737

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline