siRNA Details
| Gene Symbol: Dbndd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170975.2 | NP_001164446.2 | 2 | 329 | ||
| NM_001170976.1 | NP_001164447.2 | 2 | 329 | |||
| NM_028146.4 | NP_082422.3 | 2 | 329 | |||
| GenBank mRNA | AK013173.1 | - | 2 | 228 | ||
| AK028187.1 | - | 2 | 266 | |||
| AK163434.1 | - | 2 | 329 | |||
| BC046622.1 | - | 2 | 264 | 6389902 | ||
| BQ554936.1 | - | 2 | 236 | |||

Dbndd1
Genomic Map