This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TTC19 SIRNA Assays ›
Gene Symbol:
Ttc19
Gene Name:
tetratricopeptide repeat domain 19
Gene Aliases:
2010204O13Rik, 2810460C24Rik, AI505442
Chromosome Location:
Chr.11: 62281427 - 62316441 on Build GRCm38
UniGene ID:
Mm.489722
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 82795

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttc19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028360.2 NP_082636.3 5, 6 529
NM_029704.2 NP_083980.2 4, 5 490
GenBank mRNA AK013368.1 - 4, 5 482
AK034075.1 - 5, 6 519
AK213856.1 - 110
BC026866.1 - 3, 4 173
BC037111.1 - 4, 5 253 5370464

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109644, XM_006246530.

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial respiratory chain complex III assembly
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-29j8d:80/100.66.77.18:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline