This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ARMC8 SIRNA Assays ›
Gene Symbol:
Armc8
Gene Name:
armadillo repeat containing 8
Gene Aliases:
1200015K23Rik, Gid5, HSPC056
Chromosome Location:
Chr.9: 99478385 - 99568936 on Build GRCm38
UniGene ID:
Mm.234823
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 83182

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Armc8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166138.1 NP_001159610.1 2 341
NM_028768.3 NP_083044.2 2 341
GenBank mRNA AK004793.2 - 2 342
AK078280.1 - 2 92
BC038866.1 - 2 299 1230321

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: CX167429, AK289577, AF109184, AK296837, BC013424, and others.
This siRNA also targets the following transcripts in Rat: NM_001173354, XM_006243622, XM_006243624, XM_006243623, BC168192.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0