siRNA Details
| Gene Symbol: Cep97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159364.1 | NP_001152836.1 | 3 | 358 | ||
| NM_001159365.1 | NP_001152837.1 | 3 | 358 | |||
| NM_001159366.1 | NP_001152838.1 | 3 | 358 | |||
| NM_028815.4 | NP_083091.1 | 3 | 358 | |||
| GenBank mRNA | AK012971.2 | - | 3 | 343 | ||
| AK040079.1 | - | 3 | 365 | |||
| AK046486.1 | - | 3 | 907 | |||
| AK079884.1 | - | 3 | 326 | |||
| AK080249.1 | - | 3 | 344 | |||
| AK154885.1 | - | 3 | 337 | |||
| BC065773.1 | - | 3 | 307 | |||
| BC067049.1 | - | 3 | 358 | 30363673 | ||

Cep97
Genomic Map