siRNA Details
| Gene Symbol: Tmem43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028766.2 | NP_083042.1 | 5 | 558 | ||
| GenBank mRNA | AK004778.1 | - | 5 | 559 | ||
| AK037165.1 | - | 5 | 530 | |||
| AK050173.1 | - | 5 | 597 | |||
| AK151693.1 | - | 5 | 551 | |||
| AK151766.1 | - | 5 | 576 | |||
| AK152078.1 | - | 5 | 551 | |||
| AK168559.1 | - | 5 | 566 | |||
| BC005521.1 | - | 5 | 523 | |||
| BC024933.1 | - | 5 | 545 | 3990353 | ||

Tmem43
Genomic Map