This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SGMS2 SIRNA Assays ›
Gene Symbol:
Sgms2
Gene Name:
sphingomyelin synthase 2
Gene Aliases:
4933405A16Rik, 5133401H06Rik, AI854299
Chromosome Location:
Chr.3: 131318985 - 131491192 on Build GRCm38
UniGene ID:
Mm.273360
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 83356

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sgms2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028943.5 NP_083219.2 2 696
GenBank mRNA AK016659.1 - 2 710
BC117782.2 - 2 700 40092081

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
sphingolipid metabolic process
sphingomyelin biosynthetic process
phosphorylation
kinase activity
transferase activity
sphingomyelin synthase activity
ceramide cholinephosphotransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0