siRNA Details
| Gene Symbol: 0610040J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029554.4 | NP_083830.3 | 4 | 1539 | ||
| GenBank mRNA | AK002869.1 | - | 4 | 1540 | ||
| AK050792.1 | - | 4 | 1671 | |||
| AK159779.1 | - | 4 | 1502 | |||
| BC004797.1 | - | 4 | 1456 | 3589105 | ||

0610040J01Rik
Genomic Map