This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGS7BP SIRNA Assays ›
Gene Symbol:
Rgs7bp
Gene Name:
regulator of G-protein signalling 7 binding protein
Gene Aliases:
A930030I01Rik, AI842293, D13Bwg1146e, R7bp
Chromosome Location:
Chr.13: 104945904 - 105054930 on Build GRCm38
UniGene ID:
Mm.100348
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 84279

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rgs7bp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029879.2 NP_084155.2 6 1778
GenBank mRNA AK036240.1 - 6 1690
AK043529.1 - 3 586
BC118957.2 - 6 1330 40093423

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
negative regulation of signal transduction
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-fgnpl:80/100.66.76.214:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0