This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PARP9 SIRNA Assays ›
Gene Symbol:
Parp9
Gene Name:
poly (ADP-ribose) polymerase family, member 9
Gene Aliases:
ARTD9, AW214463, BC003281, Bagl, Bal, PARP-9
Chromosome Location:
Chr.16: 35938470 - 35972621 on Build GRCm38
UniGene ID:
Mm.49074
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 84383

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Parp9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030253.3 NP_084529.1 4 957
GenBank mRNA AK037903.1 - 4 524
AK050032.1 - 4 401
BC003281.1 - 4 399 3581841
BC010312.1 - 3 382
BC070466.1 - 4 957 30615225

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA repair
double-strand break repair
cellular response to DNA damage stimulus
regulation of response to interferon-gamma
NAD+ ADP-ribosyltransferase activity
transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-2scln:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline