This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other XLR5C SIRNA Assays ›
Gene Symbol:
Xlr5c Xlr5a Xlr5b
Gene Name:
X-linked lymphocyte-regulated 5C
X-linked lymphocyte-regulated 5A
X-linked lymphocyte-regulated 5B
Gene Aliases:
Xlr5
Xlr5d
OTTMUSG00000017646
Chromosome Location:
Chr.X: 73283361 - 73293970 on Build GRCm38
Chromosome Location:
Chr.X: 73107621 - 73119524 on Build GRCm38
Chromosome Location:
Chr.X: 73146976 - 73158886 on Build GRCm38
UniGene ID:
Mm.255790
Mm.435653
Mm.255790
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
siRNA ID 85116

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Xlr5c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031493.1 NP_113681.1 4 234
GenBank mRNA BC119509.2 - 4 234 40053468
Gene Symbol: Xlr5a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045539.2 NP_001039004.2 5 366
GenBank mRNA BC051681.1 - 5 366 30021721
Gene Symbol: Xlr5b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111293.1 NP_001104763.1 5 321

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

spermatid development
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gb242:80/100.66.79.144:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline