siRNA Details
| Gene Symbol: Emc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033146.1 | NP_149158.1 | 4, 5 | 582 | ||
| GenBank mRNA | AB054001.1 | - | 4, 5 | 628 | ||
| AK005154.1 | - | 4, 5 | 582 | |||
| AK144394.1 | - | 4, 5 | 610 | |||
| AK154475.1 | - | 4, 5 | 631 | |||
| AK180014.1 | - | 2, 3 | 159 | |||
| BC024704.1 | - | 4, 5 | 558 | 4195108 | ||

Emc9
Genomic Map