siRNA Details
| Gene Symbol: Lztfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033322.2 | NP_201579.1 | 3 | 196 | ||
| GenBank mRNA | AJ297352.1 | - | 3 | 192 | ||
| AK029671.1 | - | 3 | 228 | |||
| AK030068.1 | - | 3 | 208 | |||
| AK041091.1 | - | 3 | 229 | |||
| AK046515.1 | - | 3 | 185 | |||
| AK054003.1 | - | 3 | 202 | |||
| AK145263.1 | - | 3 | 205 | |||
| AK147312.1 | - | 2 | 140 | |||
| AK157770.1 | - | 3 | 203 | |||
| BC026840.1 | - | 3 | 196 | 4165152 | ||
| BC037045.1 | - | 3 | 196 | |||
| BC098224.1 | - | 3 | 201 | 1263280 | ||

Lztfl1
Genomic Map