This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CLEC2D SIRNA Assays ›
Gene Symbol:
Clec2d
Gene Name:
C-type lectin domain family 2, member d
Gene Aliases:
Clr-b, Clrb, Ocil
Chromosome Location:
Chr.6: 129180615 - 129186535 on Build GRCm38
UniGene ID:
Mm.197536
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 85589

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Clec2d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053109.3 NP_444339.1 3, 4 437
GenBank mRNA AF321553.1 - 3, 4 430
AF350409.1 - 3, 4 422
AY320031.1 - 3, 4 437
BC106776.1 - 3, 4 397 40040181
HQ713439.1 - 3, 4 371
HQ875062.1 - 3, 4 371

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular defense response
signal transduction
protection from natural killer cell mediated cytotoxicity
negative regulation of osteoclast differentiation
transmembrane signaling receptor activity
protein binding
carbohydrate binding
natural killer cell lectin-like receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline