siRNA Details
| Gene Symbol: Hemgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053149.2 | NP_444379.1 | 2 | 320 | ||
| GenBank mRNA | AF269248.1 | - | 2 | 320 | ||
| AK006588.1 | - | 3 | 335 | |||
| AK076587.1 | - | 3 | 305 | |||
| AK077692.1 | - | 2 | 163 | |||
| AK156536.1 | - | 2 | 248 | |||
| AK171890.1 | - | 2 | 239 | |||
| AK171906.1 | - | 2 | 369 | |||
| AK172421.1 | - | 2 | 239 | |||
| AY699986.1 | - | 2 | 162 | |||
| BC050798.1 | - | 3 | 299 | 6743819 | ||

Hemgn
Genomic Map