This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HSD17B11 SIRNA Assays ›
Gene Symbol:
Hsd17b11
Gene Name:
hydroxysteroid (17-beta) dehydrogenase 11
Gene Aliases:
Dhrs8, Pan1b, SDR2, retSDR2
Chromosome Location:
Chr.5: 103989765 - 104021796 on Build GRCm38
UniGene ID:
Mm.46019
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 85966

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hsd17b11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053262.3 NP_444492.1 2 287
GenBank mRNA AF304306.1 - 2 283
AK049355.1 - 2 331
AK154803.1 - 2 302
AK155174.1 - 2 302
AK155202.1 - 2 307
AK155327.1 - 2 307
AK158327.1 - 2 302
AK170939.1 - 2 302
AY053570.1 - 2 301
BC038340.1 - 2 287 4458725

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
steroid biosynthetic process
androgen catabolic process
metabolic process
oxidation-reduction process
estradiol 17-beta-dehydrogenase activity
steroid dehydrogenase activity
oxidoreductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0