siRNA Details
| Gene Symbol: Smco4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133214.2 | NP_573477.2 | 3 | 502 | ||
| GenBank mRNA | AF197136.1 | - | 3 | 502 | ||
| AK146267.1 | - | 3 | 545 | |||
| BC017612.1 | - | 3 | 380 | 4911985 | ||
| CO800370.1 | - | 3 | 407 | |||

Smco4
Genomic Map