This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CDC123 SIRNA Assays ›
Gene Symbol:
Cdc123
Gene Name:
cell division cycle 123
Gene Aliases:
AA959893, G431001I09Rik
Chromosome Location:
Chr.2: 5794294 - 5844968 on Build GRCm38
UniGene ID:
Mm.181490
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 87119

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdc123
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133837.4 NP_598598.1 2, 3 206
GenBank mRNA AK090122.1 - 2, 3 207
AK141463.1 - 2, 3 194
AK159208.1 - 2, 3 227
AK161246.1 - 2, 3 193
BC019753.1 - 2, 3 181 5008314
BC022972.1 - 2, 3 195 5368775
BC023821.1 - 2, 3 198 5354711
BC024787.1 - 2, 3 193 5359215

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell cycle
cell division
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline