siRNA Details
| Gene Symbol: Snapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133968.1 | NP_598729.1 | 2 | 256 | ||
| GenBank mRNA | AK004379.1 | - | 2 | 262 | ||
| AK035340.1 | - | 2 | 268 | |||
| AK075569.1 | - | 2 | 279 | |||
| AK141475.1 | - | 2 | 258 | |||
| AK146218.1 | - | 2 | 248 | |||
| AK155012.1 | - | 2 | 268 | |||
| BC011147.1 | - | 2 | 256 | 4194905 | ||

Snapc2
Genomic Map