siRNA Details
| Gene Symbol: PEX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000466.2 | NP_000457.1 | 24 | 4117 | ||
| NM_001282677.1 | NP_001269606.1 | 23 | 3946 | |||
| NM_001282678.1 | NP_001269607.1 | 24 | 4152 | |||
| XM_017012319.1 | XP_016867808.1 | 19 | 3037 | |||
| GenBank mRNA | AB008112.1 | - | 24 | 4088 | ||
| AB209355.1 | - | 12 | 5071 | |||
| AF026086.1 | - | 24 | 4081 | |||
| AF030356.1 | - | 24 | 4025 | |||
| AK292955.1 | - | 24 | 4022 | |||
| AK295686.1 | - | 21 | 3061 | |||
| BC035575.1 | - | 24 | 4059 | 5498274 | ||

PEX1
Genomic Map