siRNA Details
| Gene Symbol: Zfp286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138949.3 | NP_620399.3 | 3, 4 | 296 | ||
| GenBank mRNA | AK031547.1 | - | 3, 4 | 306 | ||
| BC038632.1 | - | 3, 4 | 288 | |||
| BC050969.1 | - | 3, 4 | 302 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp286| Gene Symbol: Zfp286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138949.3 | NP_620399.3 | 3, 4 | 296 | ||
| GenBank mRNA | AK031547.1 | - | 3, 4 | 306 | ||
| BC038632.1 | - | 3, 4 | 288 | |||
| BC050969.1 | - | 3, 4 | 302 | |||