This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other WDR36 SIRNA Assays ›
Gene Symbol:
Wdr36
Gene Name:
WD repeat domain 36
Gene Aliases:
5730444A13Rik, Ta-wdrp
Chromosome Location:
Chr.18: 32837225 - 32866420 on Build GRCm38
UniGene ID:
Mm.38816
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 88858

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wdr36
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110015.1 NP_001103485.1 3 416
NM_001110016.1 NP_001103486.1 3 416
NM_144863.4 NP_659112.2 3 416
GenBank mRNA AK041763.1 - 3 336
AK045396.1 - 3 389
AK077545.1 - 3 346
AK140019.1 - 3 341
AK155965.1 - 3 315
AK156822.1 - 417
AK171683.1 - 3 354
AK172054.1 - 3 341
AK172377.1 - 3 346
BC022132.1 - 3 388
BC025809.1 - 3 330
BC094453.1 - 3 367 5068941

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
rRNA processing
regulation of axon extension
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline