This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DIRAS1 SIRNA Assays ›
Gene Symbol:
Diras1 Diras2
Gene Name:
DIRAS family, GTP-binding RAS-like 1
DIRAS family, GTP-binding RAS-like 2
Gene Aliases:
Di-Ras1, Gbts1, RIG
2900052J15Rik, AI414999
Chromosome Location:
Chr.10: 81019589 - 81025821 on Build GRCm38
Chromosome Location:
Chr.13: 52504375 - 52531242 on Build GRCm38
UniGene ID:
Mm.44995
Mm.29362
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 89086

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Diras1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145217.2 NP_660252.1 2 240
GenBank mRNA AK081336.1 - 2 239
AY059640.1 - 1 146
BC057556.1 - 2 240 6810812
Gene Symbol: Diras2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024474.2 NP_001019645.1 2 599
GenBank mRNA AK041534.1 - 2 1042
AK136096.1 - 2 600
AK136101.1 - 2 600
BC086799.1 - 2 417 6833911

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108987, NM_001169578, XM_006253671, XM_008771489.

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
small GTPase mediated signal transduction
metabolic process
biological_process
nucleotide binding
GTPase activity
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-sp9vq:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline