siRNA Details
| Gene Symbol: Tiprl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145513.4 | NP_663488.1 | 2 | 373 | ||
| GenBank mRNA | AK049119.1 | - | 2 | 321 | ||
| AK049784.1 | - | 2 | 359 | |||
| AK075774.1 | - | 2 | 353 | |||
| BC002098.1 | - | 2 | 318 | 3590157 | ||
| BC058250.1 | - | 2 | 312 | 6315314 | ||
| CF531065.1 | - | 2 | 375 | |||

Tiprl
Genomic Map