siRNA Details
| Gene Symbol: Cyp2j13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145548.4 | NP_663523.2 | 7 | 1097 | ||
| GenBank mRNA | AK143903.1 | - | 7 | 1098 | ||
| AK143981.1 | - | 7 | 1094 | |||
| AK162358.1 | - | 1101 | ||||
| AK165405.1 | - | 7 | 1097 | |||
| AK165469.1 | - | 7 | 1075 | |||
| AK165473.1 | - | 7 | 1075 | |||
| BC016446.1 | - | 6 | 910 | 4235131 | ||
| BC024451.1 | - | 6 | 717 | |||

Cyp2j13
Genomic Map