This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM5136 SIRNA Assays ›
Gene Symbol:
Gm5136 Brcc3
Gene Name:
predicted gene 5136
BRCA1/BRCA2-containing complex, subunit 3
Gene Aliases:
C6.1AL, EG368203
C6.1A
Chromosome Location:
Chr.10: 108699044 - 108700160 on Build GRCm38
Chromosome Location:
Chr.X: 75416628 - 75455702 on Build GRCm38
UniGene ID:
Mm.452758
Mm.226957
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 90193

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm5136
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203660.2 NP_988991.1 1 169
GenBank mRNA BC120506.1 - 1 169 40129429
BC120508.1 - 1 169 40129431
Gene Symbol: Brcc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166457.1 NP_001159929.1 1 192
NM_001166459.1 NP_001159931.1 1 192
NM_145956.4 NP_666068.1 1 192
GenBank mRNA AK011876.1 - 1 109
AK028483.1 - 1 121
AK030736.1 - 1 122
AK038919.1 - 1 110
AK149844.1 - 1 124
BC021313.1 - 1 139 5026715
BC048179.1 - 1 106 6825974
S68022.1 - 1 134

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
biological_process
DNA repair
double-strand break repair
cellular response to DNA damage stimulus
response to X-ray
response to ionizing radiation
chromatin modification
G2 DNA damage checkpoint
positive regulation of DNA repair
regulation of catalytic activity
protein K63-linked deubiquitination
histone H2A K63-linked deubiquitination
molecular_function
peptidase activity
metallopeptidase activity
hydrolase activity
pyroglutamyl-peptidase activity
metal ion binding
thiol-dependent ubiquitin-specific protease activity
protein binding
enzyme regulator activity
polyubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline