This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CEP290 SIRNA Assays ›
Gene Symbol:
Cep290
Gene Name:
centrosomal protein 290
Gene Aliases:
BC004690, Nphp6, b2b1454Clo, b2b1752Clo
Chromosome Location:
Chr.10: 100487548 - 100573655 on Build GRCm38
UniGene ID:
Mm.229114
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 90513

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cep290
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146009.2 NP_666121.2 37 5087
GenBank mRNA AK029960.1 - 2 94
BC004690.1 - 3 373

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001135755.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transport
establishment or maintenance of cell polarity
protein transport
cell projection organization
regulation of cAMP metabolic process
nonmotile primary cilium assembly
cilium assembly
photoreceptor cell maintenance
positive regulation of transcription, DNA-templated
retina development in camera-type eye
cilium morphogenesis
regulation of establishment of protein localization
positive regulation of intracellular protein transport
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline