siRNA Details
| Gene Symbol: Olfr518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146306.1 | NP_666418.1 | 1 | 85 | ||
| GenBank mRNA | BC104128.1 | - | 1 | 85 | ||
| BC104129.1 | - | 1 | 85 | |||
| BC114642.1 | - | 1 | 84 | |||
| BC115846.1 | - | 1 | 84 | |||
| BC127222.1 | - | 1 | 85 | 40044607 | ||
| BC127223.1 | - | 1 | 85 | 40044609 | ||

Olfr518
Genomic Map