This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR1484 SIRNA Assays ›
Gene Symbol:
Olfr1484 Olfr1477 Olfr1487 Olfr1489
Gene Name:
olfactory receptor 1484
olfactory receptor 1477
olfactory receptor 1487
olfactory receptor 1489
Gene Aliases:
MOR202-37
MOR202-10
MOR202-18
MOR202-19
Chromosome Location:
Chr.19: 13583556 - 13586420 on Build GRCm38
Chromosome Location:
Chr.19: 13500546 - 13503292 on Build GRCm38
Chromosome Location:
Chr.19: 13619164 - 13620111 on Build GRCm38
Chromosome Location:
Chr.19: 13633113 - 13634054 on Build GRCm38
UniGene ID:
Mm.451652
Mm.271532
Mm.271479
Mm.271478
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
siRNA ID 91116

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr1484
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146291.1 NP_666403.1 1 713
Gene Symbol: Olfr1477
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146696.2 NP_666907.2 2 865
Gene Symbol: Olfr1487
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146636.1 NP_666847.1 1 713
Gene Symbol: Olfr1489
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146635.1 NP_666846.1 1 710
GenBank mRNA BC111827.1 - 1 709

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0