This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR518 SIRNA Assays ›
Gene Symbol:
Olfr518
Gene Name:
olfactory receptor 518
Gene Aliases:
MOR268-5
Chromosome Location:
Chr.7: 108880603 - 108881604 on Build GRCm38
UniGene ID:
Mm.377423
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 91130

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr518
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146306.1 NP_666418.1 1 280
GenBank mRNA BC104128.1 - 1 280
BC104129.1 - 1 280
BC114642.1 - 1 279
BC115846.1 - 1 279
BC127222.1 - 1 280 40044607
BC127223.1 - 1 280 40044609

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline