This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC29A4 SIRNA Assays ›
Gene Symbol:
Slc29a4
Gene Name:
solute carrier family 29 (nucleoside transporters), member 4
Gene Aliases:
ENT4, mPMAT
Chromosome Location:
Chr.5: 142692476 - 142722490 on Build GRCm38
UniGene ID:
Mm.125942
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 91174

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc29a4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146257.2 NP_666369.1 11 2603
GenBank mRNA BC025599.1 - 11 2547 5250967

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
monoamine transport
transmembrane transport
nucleoside transmembrane transporter activity
monoamine transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline