This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR596 SIRNA Assays ›
Gene Symbol:
Olfr596 Olfr603
Gene Name:
olfactory receptor 596
olfactory receptor 603
Gene Aliases:
ENSMUSG00000073953, Gm15117, OTTMUSG00000019046, Olfr596-ps1
MOR32-14_i, MOR32-2, Olfr603-ps1, Olfr604
Chromosome Location:
Chr.7: 103309723 - 103310661 on Build GRCm38
Chromosome Location:
Chr.7: 103383062 - 103384000 on Build GRCm38
UniGene ID:
Mm.482068
Mm.377824
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 92371

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr596
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190381.1 NP_001177310.1 1 592
Gene Symbol: Olfr603
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147070.2 NP_667281.2 1 592

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000543.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
molecular_function
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline