This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM18 SIRNA Assays ›
Gene Symbol:
Tmem18
Gene Name:
transmembrane protein 18
Gene Aliases:
BC024093
Chromosome Location:
Chr.12: 30584443 - 30591219 on Build GRCm38
UniGene ID:
Mm.34035
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 93515

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172049.2 NP_742046.2 2 214
GenBank mRNA AK133493.1 - 2 214
AK160817.1 - 2 206
AK166323.1 - 2 231
AK170496.1 - 2 197
BC024093.1 - 2 186 5116657
BY013255.1 - 2 215

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
cell migration
molecular_function
DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline