siRNA Details
| Gene Symbol: Mab21l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172295.4 | NP_758499.3 | 7 | 2910 | ||
| GenBank mRNA | AK029040.1 | - | 7 | 2649 | ||
| AK161057.1 | - | 7 | 2827 | |||
| AK166928.1 | - | 2660 | ||||
| AK172389.1 | - | 7 | 2859 | |||
| BC037703.1 | - | 7 | 2847 | 4166596 | ||

Mab21l3
Genomic Map