siRNA Details
| Gene Symbol: UGT2B15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076.3 | NP_001067.2 | 6 | 2156 | ||
| GenBank mRNA | AF180322.1 | - | 6 | 2037 | ||
| AF548389.1 | - | 6 | 2015 | |||
| U06641.1 | - | 6 | 2016 | |||
| U08854.1 | - | 6 | 2021 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT2B15| Gene Symbol: UGT2B15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076.3 | NP_001067.2 | 6 | 2156 | ||
| GenBank mRNA | AF180322.1 | - | 6 | 2037 | ||
| AF548389.1 | - | 6 | 2015 | |||
| U06641.1 | - | 6 | 2016 | |||
| U08854.1 | - | 6 | 2021 | |||