This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZDHHC9 SIRNA Assays ›
Gene Symbol:
Zdhhc9
Gene Name:
zinc finger, DHHC domain containing 9
Gene Aliases:
6430508G22, 9530098M12Rik
Chromosome Location:
Chr.X: 48171967 - 48208702 on Build GRCm38
UniGene ID:
Mm.207367
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 94066

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zdhhc9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172465.4 NP_766053.1 10 2292
GenBank mRNA AK032233.1 - 10 2274
BC042618.1 - 10 2151 4950217
BC090832.1 - 10 2110 6401763

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

peptidyl-L-cysteine S-palmitoylation
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
protein-cysteine S-palmitoyltransferase activity
Ras palmitoyltransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline