This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DPP9 SIRNA Assays ›
Gene Symbol:
Dpp9
Gene Name:
dipeptidylpeptidase 9
Gene Aliases:
6430584G11Rik, A330078I11, DPP IX, DPRP2
Chromosome Location:
Chr.17: 56186674 - 56218889 on Build GRCm38
UniGene ID:
Mm.205535
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 94496

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dpp9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172624.3 NP_766212.2 8 1072
GenBank mRNA AK039652.1 - 7 1165
AK050021.1 - 8 1014
AK050023.1 - 8 1074
AK078301.1 - 8 1020
AK131178.1 - 5 2935
AK219044.1 - 2 89
BC057631.1 - 8 1071 4191886

Back To Top

More Information


Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
aminopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline