siRNA Details
| Gene Symbol: Gramd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172768.1 | NP_766356.1 | 8 | 1106 | ||
| GenBank mRNA | AK028277.1 | - | 9 | 1001 | ||
| AK033156.1 | - | 8 | 1106 | |||
| AK122465.1 | - | 7 | 1066 | |||
| BC070451.1 | - | 7 | 940 | 30616509 | ||

Gramd1b
Genomic Map